Learn More
Abnova™ Human SYNGR2 Partial ORF (NP_004701, 168 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009144-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an integral membrane protein containing four transmembrane regions and a C-terminal cytoplasmic tail that is tyrosine phosphorylated. The exact function of this protein is unclear, but studies of a similar rat protein suggest that it may play a role in regulating membrane traffic in non-neuronal cells. The gene belongs to the synaptogyrin gene family. [provided by RefSeq]
Sequence: QRYKAGVDDFIQNYVDPTPDPNTAYASYPGASVDNYQQPPFTQNAETTEGYQPPPVYSpecifications
NP_004701 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.01kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QRYKAGVDDFIQNYVDPTPDPNTAYASYPGASVDNYQQPPFTQNAETTEGYQPPPVY | |
RUO | |
SYNGR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9144 | |
SYNGR2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC102914 | |
SYNGR2 | |
Recombinant | |
wheat germ expression system |