missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAF10 Full-length ORF (NP_006275.1, 1 a.a. - 218 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number NP_006275.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6881
Molecular Weight (g/mol) 48.1kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16133388
View Documents Promotion Details
Abnova™
H00006881-P01.25UG
25 ug
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16123388
View Documents Promotion Details
Abnova™
H00006881-P01.10UG
10 ug
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs. [provided by RefSeq]

Sequence: MSCSGSGADPEAAPASAASAPGPAPPVSAPAALPSSTAAENKASPAGTAGGPGAGAAAGGTGPLAARAGEPAERRGAAPVSAGGAAPPEGAISNGVYVLPSAANGDVKPVVSSTPLVDFLMQLEDYTPTIPDAVTGYYLNRAGFEASDPRIIRLISLAAQKFISDIANDALQHCKMKGTASGSSRSKSKDRKYTLTMEDLTPALSEYGINVKKPHYFT
Specifications

Specifications

NP_006275.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.1kDa
Glutathione Sepharose 4 Fast Flow
MSCSGSGADPEAAPASAASAPGPAPPVSAPAALPSSTAAENKASPAGTAGGPGAGAAAGGTGPLAARAGEPAERRGAAPVSAGGAAPPEGAISNGVYVLPSAANGDVKPVVSSTPLVDFLMQLEDYTPTIPDAVTGYYLNRAGFEASDPRIIRLISLAAQKFISDIANDALQHCKMKGTASGSSRSKSKDRKYTLTMEDLTPALSEYGINVKKPHYFT
RUO
TAF10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
6881
TAF10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAF2A/TAF2H/TAFII30
TAF10
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications