Learn More
Abnova™ Human TAF5L Partial ORF (NP_055224, 1 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_055224 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27097 |
Molecular Weight (g/mol) | 38.61kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188542
|
Abnova™
H00027097-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178542
|
Abnova™
H00027097-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene belongs to the WD-repeat TAF5 family of proteins. This gene encodes a protein that is a component of the PCAF histone acetylase complex. The PCAF histone acetylase complex, which is composed of more than 20 polypeptides some of which are TAFs, is required for myogenic transcription and differentiation. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors to facilitate complex assembly and transcription initiation. The encoded protein is structurally similar to one of the histone-like TAFs, TAF5. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVESpecifications
NP_055224 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.61kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAF65B | |
TAF5L | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27097 | |
TAF5L (Human) Recombinant Protein (Q01) | |
MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVE | |
RUO | |
TAF5L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |