Learn More
Abnova™ Human TBPL1 Full-length ORF (AAH17559, 1 a.a. - 186 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009519-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes. [provided by RefSeq]
Sequence: MDADSDVALDILITNVVCVFRTRCHLNLRKIALEGANVIYKRDVGKVLMKLRKPRITATIWSSGKIICTGATSEEEAKFGARRLARSLQKLGFQVIFTDFKVVNVLAVCNMPFEIRLPEFTKNNRPHASYEPELHPAVCYRIKSLRATLQIFSTGSITVTGPNVKAVATAVEQIYPFVFESRKEILSpecifications
AAH17559 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.20kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC:8389/MGC:9620/STUD/TLF/TLP/TRF2 | |
TBPL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9519 | |
TBPL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDADSDVALDILITNVVCVFRTRCHLNLRKIALEGANVIYKRDVGKVLMKLRKPRITATIWSSGKIICTGATSEEEAKFGARRLARSLQKLGFQVIFTDFKVVNVLAVCNMPFEIRLPEFTKNNRPHASYEPELHPAVCYRIKSLRATLQIFSTGSITVTGPNVKAVATAVEQIYPFVFESRKEIL | |
RUO | |
TBPL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |