Learn More
Abnova™ Human TBX21 Partial ORF (NP_037483, 400 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00030009-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is the human ortholog of mouse Tbx21/Tbet gene. Studies in mouse show that Tbx21 protein is a Th1 cell-specific transcription factor that controls the expression of the hallmark Th1 cytokine, interferon-gamma (IFNG). Expression of the human ortholog also correlates with IFNG expression in Th1 and natural killer cells, suggesting a role for this gene in initiating Th1 lineage development from naive Th precursor cells. [provided by RefSeq]
Sequence: RAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPESSDSGLGEGDSKRRRVSPYPSSGDSSSPAGAPSPFDKEAEGQFYNYFPNSpecifications
NP_037483 | |
Liquid | |
30009 | |
TBX21 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
T-PET/T-bet/TBET/TBLYM | |
TBX21 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.59kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPESSDSGLGEGDSKRRRVSPYPSSGDSSSPAGAPSPFDKEAEGQFYNYFPN | |
RUO | |
TBX21 | |
Wheat Germ (in vitro) | |
GST |