Learn More
Abnova™ Human TCEB3 Partial ORF (NP_003189, 81 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006924-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq]
Sequence: NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMYSpecifications
NP_003189 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMY | |
RUO | |
TCEB3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6924 | |
TCEB3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38760/FLJ42849/SIII/TCEB3A | |
TCEB3 | |
Recombinant | |
wheat germ expression system |