missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TCF21 Partial ORF (NP_003197, 90 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_003197 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6943 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126385
|
Abnova™
H00006943-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16116385
|
Abnova™
H00006943-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TCF21 encodes a transcription factor of the basic helix-loop-helix family. The TCF21 product is mesoderm specific, and expressed in embryonic epicardium, mesenchyme-derived tissues of lung, gut, gonad, and both mesenchymal and glomerular epithelial cells in the kidney. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: ARMRVLSKAFSRLKTTLPWVPPDTKLSKLDTLRLASSYIAHLRQILANDKYENGYIHPVNLTWPFMVAGKPESDLKEVVTASRLCGTTASpecifications
NP_003197 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POD1/bHLHa23 | |
TCF21 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6943 | |
TCF21 (Human) Recombinant Protein (Q01) | |
ARMRVLSKAFSRLKTTLPWVPPDTKLSKLDTLRLASSYIAHLRQILANDKYENGYIHPVNLTWPFMVAGKPESDLKEVVTASRLCGTTA | |
RUO | |
TCF21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |