Learn More
Abnova™ Human TCIRG1 Partial ORF (NP_006010.2, 138 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006010.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10312 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142336
|
Abnova™
H00010312-Q02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16132336
|
Abnova™
H00010312-Q02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Through alternate splicing, this gene encodes two proteins with similarity to subunits of the vacuolar ATPase (V-ATPase) but the encoded proteins seem to have different functions. V-ATPase is a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. V-ATPase is comprised of a cytosolic V1 domain and a transmembrane V0 domain. Mutations in this gene are associated with infantile malignant osteopetrosis. [provided by RefSeq]
Sequence: PQLAAAHTDGASERTPLLQAPGGPHQDLRVNFVAGAVEPHKAPALERLLWRACRGFLIASFRELEQPLEHPVTGEPATWMTFLISYWGEQIGQKIRKISpecifications
NP_006010.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6N1C/ATP6V0A3/Atp6i/OC-116kDa/OC116/OPTB1/Stv1/TIRC7/Vph1/a3 | |
TCIRG1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10312 | |
TCIRG1 (Human) Recombinant Protein (Q02) | |
PQLAAAHTDGASERTPLLQAPGGPHQDLRVNFVAGAVEPHKAPALERLLWRACRGFLIASFRELEQPLEHPVTGEPATWMTFLISYWGEQIGQKIRKI | |
RUO | |
TCIRG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |