missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TESK2 Full-length ORF (AAH33085.1, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010420-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125052

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene product is a serine/threonine protein kinase that contains an N-terminal protein kinase domain that is structurally similar to the kinase domains of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). Its overall structure is most related to the former, indicating that it belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. This gene is predominantly expressed in testis and prostate. The developmental expression pattern of the rat gene in testis suggests an important role for this gene in meitoic stages and/or early stages of spermiogenesis. [provided by RefSeq]

Sequence: MDRSKRNSIAGFPPRVERLEEFEGGGGGEGNVSQVGRVWPSSYRALISAFSRLTRLDDFTCEKIGSGFFSEVFKVRHRASGQVMALKMNTLSSNRANMLKEVQLMNRLSHPNILRFMGVCVHQGQLHALTEYINSGNLEQLLDSNLHLPWTVRVKLAYDIAVGLSYLHFKGIFHRDLTSKNCLIKRDENGYSAVVADFGLAEKIPDVSMGSEKLAVVGSPFWMAPEVLRDEPYNEKADVFSYGIILCEIIARIQADPDYLPRTEMDPKLRPSFVEIGKTLEEILSRLQEEEQERDRKLQPTARGLLEKAPGVKRLSSLDDKIPHKSPCPRRTIWLSRSQSDIFSRKPPRTVSVLDPYYRPRDGAARTPKVNPFSARQDLMGGKIKFFDLPSKSVISLVFDLDAPGPGTMPLADWQEPLAPPIRRWCSLPGSPEFLHQEACPFVGREESLSDGPPPRLSSLKYRVKEIPPFRASALPAAQAHEAMDCSILQEENGFGSRPQGTSPCPAGASEEMEVEERPAGSTPATFSTSGIGLQTQGKQDG
Specifications

Specifications

AAH33085.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TESK2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
10420
TESK2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDRSKRNSIAGFPPRVERLEEFEGGGGGEGNVSQVGRVWPSSYRALISAFSRLTRLDDFTCEKIGSGFFSEVFKVRHRASGQVMALKMNTLSSNRANMLKEVQLMNRLSHPNILRFMGVCVHQGQLHALTEYINSGNLEQLLDSNLHLPWTVRVKLAYDIAVGLSYLHFKGIFHRDLTSKNCLIKRDENGYSAVVADFGLAEKIPDVSMGSEKLAVVGSPFWMAPEVLRDEPYNEKADVFSYGIILCEIIARIQADPDYLPRTEMDPKLRPSFVEIGKTLEEILSRLQEEEQERDRKLQPTARGLLEKAPGVKRLSSLDDKIPHKSPCPRRTIWLSRSQSDIFSRKPPRTVSVLDPYYRPRDGAARTPKVNPFSARQDLMGGKIKFFDLPSKSVISLVFDLDAPGPGTMPLADWQEPLAPPIRRWCSLPGSPEFLHQEACPFVGREESLSDGPPPRLSSLKYRVKEIPPFRASALPAAQAHEAMDCSILQEENGFGSRPQGTSPCPAGASEEMEVEERPAGSTPATFSTSGIGLQTQGKQDG
RUO
TESK2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers