missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TFAP4 Full-length ORF (NP_003214.1, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007023-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136505

  • 6845.00 SEK / 25µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG (Mermod et al., 1988 [PubMed 2833704]; Hu et al., 1990 [PubMed 2123466]).[supplied by OMIM]

Sequence: MEYFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIAQQVQLQQQQEQVRLLHQEKLEREQQQLRTQLLPPPAPTHHPTVIVPAPPPPPSHHINVVTMGPSSVINSVSTSRQNLDTIVQAIQHIEGTQEKQELEEEQRRAVIVKPVRSCPEAPTSDTASDSEASDSDAMDQSREEPSGDGELP
Specifications

Specifications

NP_003214.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AP-4/bHLHc41
TFAP4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7023
TFAP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEYFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIAQQVQLQQQQEQVRLLHQEKLEREQQQLRTQLLPPPAPTHHPTVIVPAPPPPPSHHINVVTMGPSSVINSVSTSRQNLDTIVQAIQHIEGTQEKQELEEEQRRAVIVKPVRSCPEAPTSDTASDSEASDSDAMDQSREEPSGDGELP
RUO
TFAP4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers