missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TFDP3 Partial ORF (NP_057605, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051270-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16116776

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TFDP3 is a member of the DP family of proteins, which form heterodimers with E2F proteins (see E2F1; MIM 189971) and regulate transcription (Qiao et al., 2007 [PubMed 17062573]).[supplied by OMIM]

Sequence: MPQRPAASNIPVVGSPNPPSTHFASQNQHSYSSPPWAGQHNRKGEKNGMGLCRLSMKVWETVQRKGTTSCQEVVGELVAKFRAASNHASPNESAYDVKNI
Specifications

Specifications

NP_057605
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MPQRPAASNIPVVGSPNPPSTHFASQNQHSYSSPPWAGQHNRKGEKNGMGLCRLSMKVWETVQRKGTTSCQEVVGELVAKFRAASNHASPNESAYDVKNI
RUO
TFDP3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51270
TFDP3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CT30/E2F-like/HCA661/MGC161639
TFDP3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers