Läs mer
R&D Systems™ Human TGF-beta 2 Quantikine ELISA Kit
Produktkod. 18762024
Kvantitet:
1 tallrik
6 tallrikar
Produktkod. 18762024
Brand: R&D Systems™ DB250
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Human TGF-beta 2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human TGF-beta 2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 2 in cell culture supernates (125 ul), serum (125 ul), edta plasma (125 ul), heparin plasma (125 ul), citrate plasma (125 ul).
TRUSTED_SUSTAINABILITY
Specifikationer
Analysintervall 31.20 - 2000pg/mL
Analyskänslighet 7pg/mL
Konjugera HRP
Korsreaktivitet <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Produkttyp Solid Phase Sandwich ELISA
Provtyp Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificitet Natural and Recombinant Human TGF-beta 2 and natural Porcine TGF-beta 2.
Tillräckligt för 1 x 96-well Plate
Målarter Human
För användning med (applikation) ELISA
Gen-ID (Entrez) 7042
Kvantitet 1 tallrik
Provvolym Cell Culture Supernates (125uL),Serum (125uL),EDTA Plasma (125uL),Heparin Plasma (125uL),Citrate Plasma (125uL)
Förvaringskrav Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2