missing translation for 'onlineSavingsMsg'
Läs mer

Human TGF-beta RI/ALK-5 Antibody, R&D Systems™

Produktkod. 18043132
Change view
Click to view available options
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18043132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18043132 Supplier R&D Systems Supplier No. AF3025

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

TGF-beta RI/ALK-5 Polyclonal specifically detects TGF-beta RI/ALK-5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-beta RI/ALK-5
Användningsområden Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 0.1-0.3 ug/mL, Immunocytochemistry 5-15 ug/mL
Genaccessionsnr. P36897
Gene Alias AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I
Gensymboler TGFBR1
Värd art Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TGF-beta RI/ALK-5 Leu34-Glu125 Accession # P36897
Reningsmetod Affinity Purified
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7046
Testspecificitet Detects human TGF-beta RI/ALK-5 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse TGF-beta RI is observed and less than 5% cross-reactivity with recombinant human (rh) TGF-beta RII and rhTGF-beta RIII is observed.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.