Learn More
Abnova™ Human TGM1 Partial ORF (NP_000350.1, 146 a.a. - 245 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007051-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a membrane protein that catalyzes the addition of an alkyl group from an akylamine to a glutamine residue of a protein, forming an alkylglutamine in the protein. This protein alkylation leads to crosslinking of proteins and catenation of polyamines to proteins. This gene contains either one or two copies of a 22 nt repeat unit in its 3' UTR. Mutations in this gene have been associated with autosomal recessive lamellar ichthyosis (LI) and nonbullous congenital ichthyosiform erythroderma (NCIE). [provided by RefSeq]
Sequence: PFHMLLLLSRTYESSDRITLELLIGNNPEVGKGTHVIIPVGKGGSGGWKAQVVKASGQNLNLRVHTSPNAIIGKFQFTVRTQSDAGEFQLPFDPRNEIYISpecifications
NP_000350.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PFHMLLLLSRTYESSDRITLELLIGNNPEVGKGTHVIIPVGKGGSGGWKAQVVKASGQNLNLRVHTSPNAIIGKFQFTVRTQSDAGEFQLPFDPRNEIYI | |
RUO | |
TGM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7051 | |
TGM1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ICR2/KTG/LI/LI1/TGASE/TGK | |
TGM1 | |
Recombinant | |
wheat germ expression system |