Learn More
Abnova™ Human THEM4 Partial ORF (NP_444283.2, 179 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00117145-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Protein kinase B (PKB) is a major downstream target of receptor tyrosine kinases that signal via phosphatidylinositol 3-kinase. Upon cell stimulation, PKB is translocated to the plasma membrane, where it is phosphorylated in the C-terminal regulatory domain. The protein encoded by this gene negatively regulates PKB activity by inhibiting phosphorylation. Transcription of this gene is commonly downregulated in glioblastomas. [provided by RefSeq]
Sequence: NLNINYKRPIPLCSVVMINSQLDKVEGRKFFVSCNVQSVDEKTLYSEATSLFIKLNPAKSLTSpecifications
NP_444283.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.56kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NLNINYKRPIPLCSVVMINSQLDKVEGRKFFVSCNVQSVDEKTLYSEATSLFIKLNPAKSLT | |
RUO | |
THEM4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
117145 | |
THEM4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTMP/MGC29636 | |
THEM4 | |
Recombinant | |
wheat germ expression system |