missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THOC5 Full-length ORF (AAH03615.1, 1 a.a. - 683 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008563-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122592

  • 4515.00 SEK / 10µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSSESSKKRKPKVIRSDGAPAEGKRNRSDTEQEGKYYSEEAEVDLRDPGRDYELYKYTCQELQRLMAEIQDLKSRGGKDVAIEIEERRIQSCVHFMTLKKLNRLAHIRLKKGRDQTHEAKQKVDAYHLQLQNLLYEVMHLQKEITKCLEFKSKHEEIDLVSLEEFYKEAPPDISKAEVTMGDPHQQTLARLDWELEQRKRLAEKYRECLSNKEKILKEIEVKKEYLSSLQPRLNSIMQASLPVQEYLFMPFDQAHKQYETARHLPPPLYVLFVQATAYGQACDKTLSVAIEGSVDEAKALFKPPEDSQDDESDSDAEEEQTTKRRRPTLGVQLDDKRKEMLKRHPLSVMLDLKCKDDSVLHLTFYYLMNLNIMTVKAKVTTAMELITPISAGDLLSPDSVLSCLYPGDHGKKTPNPANQYQFDKVGILTLSDYVLELGHPYLWVQKLGGLHFPKEQPQQTVIADHSLSASHMETTMKLLKTRVQSRLALHKQFASLEHGIVPVTSDCQYLFPAKVVSRLVKWVTIAHEDYMELHFTKDIVDAGLAGDTNLYYMALIERGTAKLQAAVVLNPGYSSIPPIFQLCLNWKGEKTNSNDDNIRAMEGEVNVCYKELCGPWPSHQLLTNQLQRLCVLLDVYLETESHDDSVEGPKEFPQEKMCLRLFRGPSRMKPFKYNHPQGFFSHR
Specifications

Specifications

AAH03615.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
104.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C22orf19/Fmip/PK1.3/fSAP79
THOC5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8563
THOC5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSESSKKRKPKVIRSDGAPAEGKRNRSDTEQEGKYYSEEAEVDLRDPGRDYELYKYTCQELQRLMAEIQDLKSRGGKDVAIEIEERRIQSCVHFMTLKKLNRLAHIRLKKGRDQTHEAKQKVDAYHLQLQNLLYEVMHLQKEITKCLEFKSKHEEIDLVSLEEFYKEAPPDISKAEVTMGDPHQQTLARLDWELEQRKRLAEKYRECLSNKEKILKEIEVKKEYLSSLQPRLNSIMQASLPVQEYLFMPFDQAHKQYETARHLPPPLYVLFVQATAYGQACDKTLSVAIEGSVDEAKALFKPPEDSQDDESDSDAEEEQTTKRRRPTLGVQLDDKRKEMLKRHPLSVMLDLKCKDDSVLHLTFYYLMNLNIMTVKAKVTTAMELITPISAGDLLSPDSVLSCLYPGDHGKKTPNPANQYQFDKVGILTLSDYVLELGHPYLWVQKLGGLHFPKEQPQQTVIADHSLSASHMETTMKLLKTRVQSRLALHKQFASLEHGIVPVTSDCQYLFPAKVVSRLVKWVTIAHEDYMELHFTKDIVDAGLAGDTNLYYMALIERGTAKLQAAVVLNPGYSSIPPIFQLCLNWKGEKTNSNDDNIRAMEGEVNVCYKELCGPWPSHQLLTNQLQRLCVLLDVYLETESHDDSVEGPKEFPQEKMCLRLFRGPSRMKPFKYNHPQGFFSHR
RUO
THOC5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers