missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human THRA Partial ORF (AAH02728, 87 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH02728 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7067 |
Molecular Weight (g/mol) | 35.86kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136625
|
Abnova™
H00007067-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126625
|
Abnova™
H00007067-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: PTYSCKYDSCCVIDKITRNQCQLCRFKKCIAVGMAMDLVLDDSKRVAKRKLIEQNRERRRKEEMIRSLQQRPEPTPEEWDLIHIATEAHRSTSpecifications
AAH02728 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AR7/EAR7/ERB-T-1/ERBA/ERBA1/MGC000261/MGC43240/NR1A1/THRA1/THRA2/c-ERBA-1 | |
THRA | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7067 | |
THRA (Human) Recombinant Protein (Q01) | |
PTYSCKYDSCCVIDKITRNQCQLCRFKKCIAVGMAMDLVLDDSKRVAKRKLIEQNRERRRKEEMIRSLQQRPEPTPEEWDLIHIATEAHRST | |
RUO | |
THRA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |