missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Thrombopoietin R/Tpo R Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1016
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Thrombopoietin R/Tpo R Polyclonal specifically detects Thrombopoietin R/Tpo R in Human samples. It is validated for Western Blot.
Specifikationer
| Thrombopoietin R/Tpo R | |
| Polyclonal | |
| Biotin | |
| P40238 | |
| MPL | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombopoietin R/Tpo R Gln26-Tyr423 Accession # P40238 | |
| 50 μg | |
| Primary | |
| Detects human Thrombopoietin R/Tpo R in Western blots. In this format, approximately 50% cross-reactivity with recombinant mouse Tpo R is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD110, CD110 antigen, C-MPL, MPL, MPLV, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TpoR, TPO-R, TPORMPLV | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4352 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering