missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THRSP Full-length ORF (NP_003242.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007069-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133638

  • 6025.00 SEK / 25µg
Estimated Shipment: 08-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism. [provided by RefSeq]

Sequence: MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKVAGSEENGTAETEEVEDESASGELDLEAQFHLHFSSLHHILMHLTEKAQEVTRKYQEMTGQVW
Specifications

Specifications

NP_003242.1
Liquid
7069
THRSP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKVAGSEENGTAETEEVEDESASGELDLEAQFHLHFSSLHHILMHLTEKAQEVTRKYQEMTGQVW
RUO
THRSP
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC21659/S14/SPOT14
THRSP
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers