missing translation for 'onlineSavingsMsg'
Läs mer

ACROBiosystems Human TL1A / TNFSF15 Protein, His Tag (MALS verified)
GREENER_CHOICE

Produktkod. 17276580
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17276580 100 μg
17286580 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17276580 Leverantör ACROBiosystems Leverantörsnummer TLAH5243100ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

TNF-like cytokine 1A (TL1A) and its receptors, death receptor 3 (DR3) and decoy receptor 3 (DcR3) are members of the TNF and TNF receptor superfamilies of proteins, respectively. Binding of APC-derived TL1A to lymphocytic DR3 provides co-stimulatory signals for activated lymphocytes. DR3 signaling affects not only the proliferative activity of and cytokine production by effector lymphocytes, but also critically influences the development and suppressive function of regulatory T-cells. Whereas, DcR3 restricts the function of the TL1A/DR3 complex: attenuating T-cell activation and downregulating the secretion of pro-inflammatory cytokines. Together with DR3 and DcR3, TL1A constitutes a cytokine system that actively interferes with the regulation of immune responses.

This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.4 kDa. The protein migrates as 26 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifikationer

Tillträdesnummer NP_005109.2
Molekylvikt (g/mol) 22.4 kDa
Kvantitet 100 μg
Källa Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1).
Immunogen Leu 72- Leu 251
Förvaringskrav -20°C
Gene Alias TL1A, VEGI-251, TNFSF15
Vanligt namn TL1A
Konjugera Unconjugated
Art Human
Protein Tag His Tag
Buffert PBS, pH7.4
Endotoxinnivå Less than 1.0 EU / μg by the LAL method.
Uttryckssystem HEK293
Form Powder
Protein subtyp Other Recombinant Proteins
Renhet eller kvalitetsklass >90% as determined by SDS-PAGE.
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.