missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TLN1 Partial ORF (AAH42923.1, 86 a.a. - 1430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007094-Q06.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16024895

  • 4125.00 SEK / 10µg
Estimated Shipment: 29-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a cytoskeletal protein that is concentrated in areas of cell-substratum and cell-cell contacts. The encoded protein plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. It codistributes with integrins in the cell surface membrane in order to assist in the attachment of adherent cells to extracellular matrices and of lymphocytes to other cells. The N-terminus of this protein contains elements for localization to cell-extracellular matrix junctions. The C-terminus contains binding sites for proteins such as beta-1-integrin, actin, and vinculin. [provided by RefSeq]

Sequence: RPLKIRMLDGTVKTIMVDDSKTVTDMLMTICARIGITNHDEYSLVRELMEEKKEEITGTLRKDKTLLRDEKKMEKLKQKLHTDDELNWLDHGRTLREQGVEEHETLLLRRKFFYSDQNVDSRDPVQLNLLYVQARDDILNGSHPVSFDKACEFAGFQCQIQFGPHNEQKHKAGFLDLKDFLPKEYVKQKGERKIFQAHKNCGQMSEIEAKVRYVKLARSLKTYGVSFFLVKEKMKGKNKLVPRLLGITKECVMRVDEKTKEVIQEWNLTNIKRWAASPKSFTLDFGDYQDGYYSVQTTEGEQIAQLIAGYIDIILKKKKSKDHFGLEGDEESTMLEDSVSPKKSTVLQQQYNRVGKVEHGSVALPAIMRSGASGPENFQVGSMPPAQQQITSGQMHRGHMPPLTSAQQALTGTINSSMQAVQAAQATLDDFDTLPPLGQDAASKAWRKNKMDESKHEIHSQVDAITAGTASVVNLTAGDPAETDYTAVGCAVTTISSNLTEMSRGVKLLAALLEDEGGSGRPLLQAAKGLAGAVSELLRSAQPASAEPRQNLLQAAGNVGQASGELLQQIGESDTDPHFQDALMQLAKAVASAAAALVLKAKSVAQRTEDSGLQTQVIAAATQCALSTSQLVACTKVVAPTISSPVCQEQLVEAGRLVAKAVEGCVSASQAATEDGQLLRGVGAAATAVTQALNELLQHVKAHATGAGPAGRYDQATDTILTVTENIFSSMGDAGEMVRQARILAQATSDLVNAIKADAEGESDLENSRKLLSAAKILADATAKMVEAAKGAAAHPDSEEQQQRLREAAEGLRMATNAAAQNAIKKKLVQRLEHAAKQAAASATQTIAAAQHAASTPKASAGPQPLLVQSCKAVAEQIPLLVQGVRGSQAQPDSPSAQLALIAASQSFLQPGGKMVAAAKASVPTIQDQASAMQLSQCAKNLGTALAELRTAAQKAQEACGPLEMDSALSVVQNLEKDLQEVKAAARDGKLKPLPGETMEKCTQDLGNSTKAVSSAIAQLLGEVAQGNENYAGIAARDVAGGLRSLAQAARGVAALTSDPAVQAIVLDTASDVLDKASSLIEEAKKAAGHPGDPESQQRLAQVAKAVTQALNRCVSCLPGQRDVDNALRAVGDASKRLLSDSLPPSTGTFQEAQSRLNEAAAGLNQAATELVQASRGTPQDLARASGRFGQDFSTFLEAGVEMAGQAPSQEDRAQVVSNLKGISMSSSKLLLAAKALSTDPAAPNLKSQLAAAARAVTDSINQLITMCTQQAPGQKECDNALRELETVRELLENPVQPINDMSYFGCLDSVMENSKVLGEAMTGISQNAKNGNLPEFGDAISTAS
Specifications

Specifications

AAH42923.1
Liquid
7094
TLN1 (Human) Recombinant Protein (Q06)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ILWEQ/KIAA1027/TLN
TLN1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
173.58kDa
12.5% SDS-PAGE Stained with Coomassie Blue
RPLKIRMLDGTVKTIMVDDSKTVTDMLMTICARIGITNHDEYSLVRELMEEKKEEITGTLRKDKTLLRDEKKMEKLKQKLHTDDELNWLDHGRTLREQGVEEHETLLLRRKFFYSDQNVDSRDPVQLNLLYVQARDDILNGSHPVSFDKACEFAGFQCQIQFGPHNEQKHKAGFLDLKDFLPKEYVKQKGERKIFQAHKNCGQMSEIEAKVRYVKLARSLKTYGVSFFLVKEKMKGKNKLVPRLLGITKECVMRVDEKTKEVIQEWNLTNIKRWAASPKSFTLDFGDYQDGYYSVQTTEGEQIAQLIAGYIDIILKKKKSKDHFGLEGDEESTMLEDSVSPKKSTVLQQQYNRVGKVEHGSVALPAIMRSGASGPENFQVGSMPPAQQQITSGQMHRGHMPPLTSAQQALTGTINSSMQAVQAAQATLDDFDTLPPLGQDAASKAWRKNKMDESKHEIHSQVDAITAGTASVVNLTAGDPAETDYTAVGCAVTTISSNLTEMSRGVKLLAALLEDEGGSGRPLLQAAKGLAGAVSELLRSAQPASAEPRQNLLQAAGNVGQASGELLQQIGESDTDPHFQDALMQLAKAVASAAAALVLKAKSVAQRTEDSGLQTQVIAAATQCALSTSQLVACTKVVAPTISSPVCQEQLVEAGRLVAKAVEGCVSASQAATEDGQLLRGVGAAATAVTQALNELLQHVKAHATGAGPAGRYDQATDTILTVTENIFSSMGDAGEMVRQARILAQATSDLVNAIKADAEGESDLENSRKLLSAAKILADATAKMVEAAKGAAAHPDSEEQQQRLREAAEGLRMATNAAAQNAIKKKLVQRLEHAAKQAAASATQTIAAAQHAASTPKASAGPQPLLVQSCKAVAEQIPLLVQGVRGSQAQPDSPSAQLALIAASQSFLQPGGKMVAAAKASVPTIQDQASAMQLSQCAKNLGTALAELRTAAQKAQEACGPLEMDSALSVVQNLEKDLQEVKAAARDGKLKPLPGETMEKCTQDLGNSTKAVSSAIAQLLGEVAQGNENYAGIAARDVAGGLRSLAQAARGVAALTSDPAVQAIVLDTASDVLDKASSLIEEAKKAAGHPGDPESQQRLAQVAKAVTQALNRCVSCLPGQRDVDNALRAVGDASKRLLSDSLPPSTGTFQEAQSRLNEAAAGLNQAATELVQASRGTPQDLARASGRFGQDFSTFLEAGVEMAGQAPSQEDRAQVVSNLKGISMSSSKLLLAAKALSTDPAAPNLKSQLAAAARAVTDSINQLITMCTQQAPGQKECDNALRELETVRELLENPVQPINDMSYFGCLDSVMENSKVLGEAMTGISQNAKNGNLPEFGDAISTAS
RUO
TLN1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers