Learn More
Abnova™ Human TM4SF4 Full-length ORF (NP_004608.1, 1 a.a. - 202 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007104-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that can regulate cell proliferation. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq]
Sequence: MCTGGCARCLGGTLIPLAFFGFLANILLFFPGGKVIDDNDHLSQEIWFFGGILGSGVLMIFPALVFLGLKNNDCCGCCGNEGCGKRFAMFTSTIFAVVGFLGAGYSFIISAISINKGPKCLMANSTWGYPFHDGDYLNDEALWNKCREPLNVVPWNLTLFSILLVVGGIQMVLCAIQVVNGLLGTLCGDCQCCGCCGGDGPVSpecifications
NP_004608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31015/ILTMP/il-TMP | |
TM4SF4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7104 | |
TM4SF4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCTGGCARCLGGTLIPLAFFGFLANILLFFPGGKVIDDNDHLSQEIWFFGGILGSGVLMIFPALVFLGLKNNDCCGCCGNEGCGKRFAMFTSTIFAVVGFLGAGYSFIISAISINKGPKCLMANSTWGYPFHDGDYLNDEALWNKCREPLNVVPWNLTLFSILLVVGGIQMVLCAIQVVNGLLGTLCGDCQCCGCCGGDGPV | |
RUO | |
TM4SF4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |