missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TMCO2 Full-length ORF (NP_001008740.1, 1 a.a. - 182 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00127391-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MSTSSSSSWDNLLESLSLSTVWNWIQASFLGETSAPQQTSLGLLDNLAPAVQIILRISFLILLGIGIYALWKRSIQSIQKTLLFVITLYKLYKKGSHIFEALLANPEGSGLRIQDNNNLFLSLGLQEKILKKLKTVENKMKNLEGIIVAQKPATKRDCSSEPYCSCSDCQSPLSTSGFTSPISpecifications
NP_001008740.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC131705/dJ39G22.2 | |
TMCO2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
127391 | |
TMCO2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTSSSSSWDNLLESLSLSTVWNWIQASFLGETSAPQQTSLGLLDNLAPAVQIILRISFLILLGIGIYALWKRSIQSIQKTLLFVITLYKLYKKGSHIFEALLANPEGSGLRIQDNNNLFLSLGLQEKILKKLKTVENKMKNLEGIIVAQKPATKRDCSSEPYCSCSDCQSPLSTSGFTSPI | |
RUO | |
TMCO2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |