missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human TMEM253 (aa 78-99) Control Fragment Recombinant Protein

Produktkod. 30206952
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30206952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30206952 Leverantör Invitrogen™ Leverantörsnummer RP104042
 Begränsad artikel
Beräknad leverans: 15-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64200. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRUSTED_SUSTAINABILITY

Specifica

Tillträdesnummer P0C7T8
Koncentration ≥5.0 mg/mL
För användning med (applikation) Neutralization, Control
Formulering PBS, 1M urea with no preservative; pH 7.4
Gen-ID (Entrez) 643382
Namn Human TMEM253 (aa 78-99) Control Fragment
pH-intervall 7.4
Reningsmetod Purified
Kvantitet 100 μL
Förvaringskrav -20°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias C14orf176; C14orf95; NCRNA00220; TMEM253; Transmembrane protein 253; transmembrane protein C14orf176
Vanligt namn TMEM253
Gensymbol TMEM253
Produkttyp Protein
Konjugera Unconjugated
Art Human
Rekombinant Recombinant
Protein Tag His-ABP-tag
Sekvens GTVTLELRRAPRLWKVRAMMIF
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Uttryckssystem E. coli
Form Liquid
Renhet eller kvalitetsklass >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.