Learn More
Abnova™ Human TMEM68 Full-length ORF (AAH20835.1, 1 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00137695-P01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16128103
  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
Sequence: MIDKNQTCGVGQDSVPYMICLIHILEEWFGVEQLEDYLNFANYLLWVFTPLILLILPYFTIFLLYLTIIFLHIYKRKNVLKEAYSHNLWDGARKTVATLWDGHAAVWHGKQGYFHLCVAIHVCCIGTVLPFHFID
Specifications
Specifications
AAH20835.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32370/MGC87778
TMEM68
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
137695
TMEM68 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIDKNQTCGVGQDSVPYMICLIHILEEWFGVEQLEDYLNFANYLLWVFTPLILLILPYFTIFLLYLTIIFLHIYKRKNVLKEAYSHNLWDGARKTVATLWDGHAAVWHGKQGYFHLCVAIHVCCIGTVLPFHFID
RUO
TMEM68