Learn More
Abcam Human TMPRSS2 knockout Caco-2 cell line
Product Code. 32139898 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
Unit Size:
Each
Product Code. Quantity Unit Size
32139898 1,000,000 Cells Each
1 options
Product Code. 32139898 Supplier Abcam Supplier No. ab2737371000000Cells_
 Restricted Item
Beräknad leverans: 27-11-2026
TMPRSS2 KO cell line available to order. KO validated by Western blot. Knockout achieved by using CRISPR/Cas9, Homozygous: 77bp deletion in exon 3.
TMPRSS2 KO cell line available to order. KO validated by Western blot. Knockout achieved by using CRISPR/Cas9, Homozygous: 77bp deletion in exon 3.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells
Regulatory Status RUO