Learn More
Abnova™ Human TMSB4X Full-length ORF (NP_066932, 1 a.a. - 44 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007114-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y. [provided by RefSeq]
Sequence: MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGESSpecifications
NP_066932 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.58kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FX/PTMB4/TB4X/TMSB4 | |
TMSB4X | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7114 | |
TMSB4X (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGES | |
RUO | |
TMSB4X | |
Wheat Germ (in vitro) | |
GST | |
Liquid |