missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human TNF-alpha Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 24 publications
Brand: R&D Systems BAF210
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TNF-alpha Polyclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).
Specifikationer
| TNF-alpha | |
| Polyclonal | |
| Biotin | |
| P01375 | |
| TNF | |
| E. coli-derived recombinant human TNF-alpha (R&D Systems, Catalog # 210-TA) Val77-Leu233 Accession # P01375 | |
| 50 μg | |
| Primary | |
| Detects human TNF-alpha in ELISAs and Western blots. In sandwich immunoassays, less than 0.04% cross-reactivity with recombinant human (rh) TNF-beta , recombinant mouse (rm) TNF-alpha , recombinant rat TNF-alpha , recombinant porcine TNF-alpha , rhsTNF RII, rmsTNF RI, and rmsTNF RII is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7124 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering