Läs mer
R&D Systems™ Human TNF RII/TNFRSF1B (AAA36755) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC1338
Encompass
Produktkod. 15761325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human TNF RII/TNFRSF1B (AAA36755) VersaClone cDNA clone contains the complete ORF for TNF RII/TNFRSF1B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TNF RII/TNFRSF1B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature
Specifikationer
Specifikationer
TNF RII/TNFRSF1B
10 μg at 0.2 μg/μL
1399bp
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Human
AAA36755
cDNA is provided in 10mM Tris-Cl, pH 8.11
7133
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.
For Reasearch Use Only. Not for use in Humans.