Learn More
Abnova™ Human TNNI2 Partial ORF (NP_003273.1, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007136-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a fast-twitch skeletal muscle protein, a member of the troponin I gene family, and a component of the troponin complex including troponin T, troponin C and troponin I subunits. The troponin complex, along with tropomyosin, is responsible for the calcium-dependent regulation of striated muscle contraction. Mouse studies show that this component is also present in vascular smooth muscle and may play a role in regulation of smooth muscle function. In addition to muscle tissues, this protein is found in corneal epithelium, cartilage where it is an inhibitor of angiogenesis to inhibit tumor growth and metastasis, and mammary gland where it functions as a co-activator of estrogen receptor-related receptor alpha. This protein also suppresses tumor growth in human ovarian carcinoma. Mutations in this gene cause myopathy and distal arthrogryposis type 2B. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: GDEEKRNRAITARRQHLKSVMLQIAATELEKEESRREAEKQNYLAEHCPPLHIPGSMSEVQELCKQLHAKIDAAEEEKYDMEVRVQKTSKELEDMNQKLSpecifications
NP_003273.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GDEEKRNRAITARRQHLKSVMLQIAATELEKEESRREAEKQNYLAEHCPPLHIPGSMSEVQELCKQLHAKIDAAEEEKYDMEVRVQKTSKELEDMNQKL | |
RUO | |
TNNI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7136 | |
TNNI2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMCD2B/DA2B/FSSV | |
TNNI2 | |
Recombinant | |
wheat germ expression system |