missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human TOP2B (aa 1485-1622) Control Fragment Recombinant Protein

Produktkod. 30201251
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30201251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30201251 Leverantör Invitrogen™ Leverantörsnummer RP104901
 Begränsad artikel
Beräknad leverans: 02-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111315. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TOP2B (DNA Topoisomerase II Beta) is a Protein Coding gene. This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Tillträdesnummer Q02880
Koncentration ≥5.0 mg/mL
För användning med (applikation) Neutralization, Control
Formulering PBS, 1M urea with no preservative; pH 7.4
Gen-ID (Entrez) 7155
Namn Human TOP2B (aa 1485-1622) Control Fragment
pH-intervall 7.4
Reningsmetod Purified
Kvantitet 100 μL
Förvaringskrav -20°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias antigen MLAA-44; D230016L12Rik; DNA topoisomerase 2-beta; DNA topoisomerase II beta; DNA topoisomerase II, 180 kD; DNA topoisomerase II, beta isozyme; Top-2; Top2b; top2beta; TOPIIB; topo II beta; topoisomerase (DNA) II beta; topoisomerase (DNA) II beta 180kDa; topoisomerase II beta; topoisomerase IIb; U937 associated antigen
Vanligt namn TOP2B
Gensymbol TOP2B
Produkttyp Protein
Konjugera Unconjugated
Art Human
Rekombinant Recombinant
Protein Tag His-ABP-tag
Sekvens DKVPSKTVAAKKGKPSSDTVPKPKRAPKQKKVVEAVNSDSDSEFGIPKKTTTPKGKGRGAKKRKASGSENEGDYNPGRKTSKTTSKKPKKTSFDQDSDVDIFPSDFPTEPPSLPRTGRARKEVKYFAESDEEEDDVDF
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Uttryckssystem E. coli
Form Liquid
Renhet eller kvalitetsklass >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.