missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TOR3A Partial ORF (NP_071766, 298 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064222-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179576

  • 6845.00 SEK / 25µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: TMEHLEPHLQAEIVETIDNGFGHSRLVKENLIDYFIPFLPLEYRHVRLCARDAFLSQELLYKEETLDEIAQMMVYVPKEEQLFSSQGCKSISQRINYFLS
Specifications

Specifications

NP_071766
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TMEHLEPHLQAEIVETIDNGFGHSRLVKENLIDYFIPFLPLEYRHVRLCARDAFLSQELLYKEETLDEIAQMMVYVPKEEQLFSSQGCKSISQRINYFLS
RUO
TOR3A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
64222
TOR3A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADIR/ADIR2/FLJ22345/MGC111104
TOR3A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers