missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM16 Full-length ORF (NP_006461.3, 1 a.a. - 564 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010626-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16185442

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined. [provided by RefSeq]

Sequence: MAELDLMAPGPLPRATAQPPAPLSPDSGSPSPDSGSASPVEEEDVGSSEKLGRETEEQDSDSAEQGDPAGEGKEVLCDFCLDDTRRVKAVKSCLTCMVNYCEEHLQPHQVNIKLQSHLLTEPVKDHNWRYCPAHHSPLSAFCCPDQQCICQDCCQEHSGHTIVSLDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMAAISNTVQFLEEYCKFKNTEDITFPSVYVGLKDKLSGIRKVITESTVHLIQLLENYKKKLQEFSKEEEYDIRTQVSAVVQRKYWTSKPEPSTREQFLQYAYDITFDPDTAHKYLRLQEENRKVTNTTPWEHPYPDLPSRFLHWRQVLSQQSLYLHRYYFEVEIFGAGTYVGLTCKGIDRKGEERNSCISGNNFSWSLQWNGKEFTAWYSDMETPLKAGPFRRLGVYIDFPGGILSFYGVEYDTMTLVHKFACKFSEPVYAAFWLSKKENAIRIVDLGEEPEKPAPSLVGTAP
Specifications

Specifications

NP_006461.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EBBP
TRIM16
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10626
TRIM16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAELDLMAPGPLPRATAQPPAPLSPDSGSPSPDSGSASPVEEEDVGSSEKLGRETEEQDSDSAEQGDPAGEGKEVLCDFCLDDTRRVKAVKSCLTCMVNYCEEHLQPHQVNIKLQSHLLTEPVKDHNWRYCPAHHSPLSAFCCPDQQCICQDCCQEHSGHTIVSLDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMAAISNTVQFLEEYCKFKNTEDITFPSVYVGLKDKLSGIRKVITESTVHLIQLLENYKKKLQEFSKEEEYDIRTQVSAVVQRKYWTSKPEPSTREQFLQYAYDITFDPDTAHKYLRLQEENRKVTNTTPWEHPYPDLPSRFLHWRQVLSQQSLYLHRYYFEVEIFGAGTYVGLTCKGIDRKGEERNSCISGNNFSWSLQWNGKEFTAWYSDMETPLKAGPFRRLGVYIDFPGGILSFYGVEYDTMTLVHKFACKFSEPVYAAFWLSKKENAIRIVDLGEEPEKPAPSLVGTAP
RUO
TRIM16
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers