Learn More
Abnova™ Human TRIM23 Partial ORF (AAH22510, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH22510 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 373 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131781
|
Abnova™
H00000373-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16121781
|
Abnova™
H00000373-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein is also a member of the ADP ribosylation factor family of guanine nucleotide-binding family of proteins. Its carboxy terminus contains an ADP-ribosylation factor domain and a guanine nucleotide binding site, while the amino terminus contains a GTPase activating protein domain which acts on the guanine nucleotide binding site. The protein localizes to lysosomes and the Golgi apparatus. It plays a role in the formation of intracellular transport vesicles, their movement from one compartment to another, and phopholipase D activation. Three alternatively spliced transcript variants for this gene have been described. [provided by RefSeq]
Sequence: MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQYSpecifications
AAH22510 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARD1/ARFD1/RNF46 | |
TRIM23 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
373 | |
TRIM23 (Human) Recombinant Protein (Q01) | |
MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQY | |
RUO | |
TRIM23 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |