Learn More
Abnova™ Human TRIM37 Partial ORF (NP_056109, 865 a.a. - 964 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_056109 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4591 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178021
|
Abnova™
H00004591-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16168021
|
Abnova™
H00004591-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the tripartite motif (TRIM) family, whose members are involved in diverse cellular functions such as developmental patterning and oncogenesis. The TRIM motif includes zinc-binding domains, a RING finger region, a B-box motif and a coiled-coil domain. The RING finger and B-box domains chelate zinc and might be involved in protein-protein and/or protein-nucleic acid interactions. The gene mutations are associated with mulibrey (muscle-liver-brain-eye) nanism, an autosomal recessive disorder that involves several tissues of mesodermal origin. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]
Sequence: GHLEGLQMTDLENNSETGELQPVLPEGASAAPEEGMSSDSDIECDTENEEQEEHTSVGGFHDSFMVMTQPPDEDTHSSFPDGEQIGPEDLSFNTDENSGRSpecifications
NP_056109 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0898/MUL/POB1/TEF3 | |
TRIM37 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4591 | |
TRIM37 (Human) Recombinant Protein (Q01) | |
GHLEGLQMTDLENNSETGELQPVLPEGASAAPEEGMSSDSDIECDTENEEQEEHTSVGGFHDSFMVMTQPPDEDTHSSFPDGEQIGPEDLSFNTDENSGR | |
RUO | |
TRIM37 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |