Learn More
Abnova™ Human TRIM54 Partial ORF (NP_115935, 186 a.a. - 254 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057159-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: PILASQNTKIIDSELSDGIAMLVAGNDRVQAVITQMEEVCQTIEDNSRRQKQLLNQRFESLCAVLEERKSpecifications
NP_115935 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PILASQNTKIIDSELSDGIAMLVAGNDRVQAVITQMEEVCQTIEDNSRRQKQLLNQRFESLCAVLEERK | |
RUO | |
TRIM54 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57159 | |
TRIM54 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MURF/MURF-3/RNF30 | |
TRIM54 | |
Recombinant | |
wheat germ expression system |