missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRIM65 Partial ORF (NP_775818, 419 a.a. - 517 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00201292-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: GPCSWGLCVQEDSLQAWHNGEAQRLPGVSGRLLGMDLDLASGCLTFYSLEPQTQPLYTFHALFNQPLTPVFWLLEGRTLTLCHQPGAVFPLGPQEEVLSSpecifications
NP_775818 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GPCSWGLCVQEDSLQAWHNGEAQRLPGVSGRLLGMDLDLASGCLTFYSLEPQTQPLYTFHALFNQPLTPVFWLLEGRTLTLCHQPGAVFPLGPQEEVLS | |
RUO | |
TRIM65 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
201292 | |
TRIM65 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
4732463G12Rik | |
TRIM65 | |
Recombinant | |
wheat germ expression system |