missing translation for 'onlineSavingsMsg'
Läs mer

Human TROP-2 PE-conjugated Antibody, R&D Systems™

Produktkod. 18105571
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18105571 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18105571 Leverantör R&D Systems Leverantörsnummer FAB650P

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TROP-2
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona 77220
Konjugera R-PE
Utspädning Flow Cytometry 10 uL/10^6 cells
Formulering Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Genaccessionsnr. P09758
Gene Alias Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, gp50, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, T16, TACSTD2, TROP2, TROP2GA7331, tumor-associated calcium signal transducer 2
Gensymboler TACSTD2
Värd art Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TROP-2 His27-Thr274 Accession # P09758
Reningsmetod Protein A or G purified from hybridoma culture supernatant
Kvantitet 100 Tests
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 4070
Testspecificitet Detects human TROP-2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) VCAM-1 or rhICAM-1 is observed.
Målarter Human
Innehåll och lagring Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotyp IgG2a
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.