missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRPC4AP Partial ORF (NP_056453, 341 a.a. - 451 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026133-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: VANEESEHNQASIVFPPPGASEENGLPHTSARTQLPQSMKIMHEIMYKLEVLYVLCVLLMGRQRNQVHRMIAEFKLIPGLNNLFDKLIWRKHSASALVLHGHNQNCDCSPDSpecifications
NP_056453 | |
Liquid | |
26133 | |
TRPC4AP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf188/TRRP4AP/TRUSS | |
TRPC4AP | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VANEESEHNQASIVFPPPGASEENGLPHTSARTQLPQSMKIMHEIMYKLEVLYVLCVLLMGRQRNQVHRMIAEFKLIPGLNNLFDKLIWRKHSASALVLHGHNQNCDCSPD | |
RUO | |
TRPC4AP | |
Wheat Germ (in vitro) | |
GST |