Learn More
Abnova™ Human TRPV3 Partial ORF (NP_659505, 681 a.a. - 790 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00162514-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene product belongs to a family of nonselective cation channels that function in a variety of processes, including temperature sensation and vasoregulation. The thermosensitive members of this family are expressed in subsets of sensory neurons that terminate in the skin, and are activated at distinct physiological temperatures. This channel is activated at temperatures between 22 and 40 degrees C. This gene lies in close proximity to another family member (TRPV1) gene on chromosome 17, and the two encoded proteins are thought to associate with each other to form heteromeric channels. [provided by RefSeq]
Sequence: VENVSKESERIWRLQRARTILEFEKMLPEWLRSRFRMGELCKVAEDDFRLCLRINEVKWTEWKTHVSFLNEDPGPVRRTDFNKIQDSSRNNSKTTLNAFEEVEEFPETSVSpecifications
NP_659505 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VENVSKESERIWRLQRARTILEFEKMLPEWLRSRFRMGELCKVAEDDFRLCLRINEVKWTEWKTHVSFLNEDPGPVRRTDFNKIQDSSRNNSKTTLNAFEEVEEFPETSV | |
RUO | |
TRPV3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
162514 | |
TRPV3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VRL3 | |
TRPV3 | |
Recombinant | |
wheat germ expression system |