missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRUB1 Full-length ORF (NP_631908.1, 1 a.a. - 349 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00142940-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138293

  • 6845.00 SEK / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 [PubMed 12736709]).[supplied by OMIM]

Sequence: MAASEAAVVSSPSLKTDTSPVLETAGTVAAMAATPSARAAAAVVAAAARTGSEARVSKAALATKLLSLSGVFAVHKPKGPTSAELLNRLKEKLLAEAGMPSPEWTKRKKQTLKIGHGGTLDSAARGVLVVGIGSGTKMLTSMLSGSKRYTAIGELGKATDTLDSTGRVTEEKPYDKITQEDIEGILQKFTGNIMQVPPLYSALKKDGQRLSTLMKRGEVVEAKPARPVTVYSISLQKFQPPFFTLDVECGGGFYIRSLVSDIGKELSSCANVLELTRTKQGPFTLEEHALPEDKWTIDDIAQSLEHCSSLFPAELALKKSKPESNEQVLSCEYITLNEPKREDDVIKTC
Specifications

Specifications

NP_631908.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PUS4
TRUB1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
142940
TRUB1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAASEAAVVSSPSLKTDTSPVLETAGTVAAMAATPSARAAAAVVAAAARTGSEARVSKAALATKLLSLSGVFAVHKPKGPTSAELLNRLKEKLLAEAGMPSPEWTKRKKQTLKIGHGGTLDSAARGVLVVGIGSGTKMLTSMLSGSKRYTAIGELGKATDTLDSTGRVTEEKPYDKITQEDIEGILQKFTGNIMQVPPLYSALKKDGQRLSTLMKRGEVVEAKPARPVTVYSISLQKFQPPFFTLDVECGGGFYIRSLVSDIGKELSSCANVLELTRTKQGPFTLEEHALPEDKWTIDDIAQSLEHCSSLFPAELALKKSKPESNEQVLSCEYITLNEPKREDDVIKTC
RUO
TRUB1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers