missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Human TSPAN13 (NP_055214) VersaClone cDNA

Produktkod. 18795179
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18795179 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18795179 Leverantör R&D Systems™ Leverantörsnummer RDC2421
  • ONLINE EXCLUSIVE PRICE
    3500.00 SEK
    3309.60 SEK / 10µg
    Save 190.40 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 19-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

R&D Systems™ Human TSPAN13 (NP_055214) VersaClone cDNA is a member of the transmembrane 4 superfamily, also known as the tetraspanin family.

The Human TSPAN13 (NP_055214) VersaClone cDNA clone contains the complete ORF for TSPAN13, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TSPAN13 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifikationer

Gen hTSPAN13
Tillträdesnummer NP_055214
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Infoga typ 628bp
pH 8.5
Gen-ID (Entrez) 27075
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Human
Kvantitet 10 μg
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.