missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TUSC3 Full-length ORF (NP_839952.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_839952.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7991 |
Molecular Weight (g/mol) | 66kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182232
|
Abnova™
H00007991-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16192232
|
Abnova™
H00007991-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a candidate tumor suppressor gene. It is located within a homozygously deleted region of a metastatic prostate cancer. The gene is expressed in most nonlymphoid human tissues including prostate, lung, liver, and colon. Expression was also detected in many epithelial tumor cell lines. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MGARGAPSRRRQAGRRLRYLPTGSFPFLLLLLLLCIQLGGGQKKKENLLAEKVEQLMEWSSRRSIFRMNGDKFRKFIKAPPRNYSMIVMFTALQPQRQCSVCRQANEEYQILANSWRYSSAFCNKLFFSMVDYDEGTDVFQQLNMNSAPTFMHFPPKGRPKRADTFDLQRIGFAAEQLAKWIADRTDVHIRVFRPPNYSGTIALALLVSLVGGLLYLRRNNLEFIYNKTGWAMVSLCIVFAMTSGQMWNHIRGPPYAHKNPHNGQVSYIHGSSQAQFVAESHIILVLNAAITMGMVLLNEAATSKGDVGKRRIICLVGLGLVVFFFSFLLSIFRSKYHGYPYSFLIKSpecifications
NP_839952.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGARGAPSRRRQAGRRLRYLPTGSFPFLLLLLLLCIQLGGGQKKKENLLAEKVEQLMEWSSRRSIFRMNGDKFRKFIKAPPRNYSMIVMFTALQPQRQCSVCRQANEEYQILANSWRYSSAFCNKLFFSMVDYDEGTDVFQQLNMNSAPTFMHFPPKGRPKRADTFDLQRIGFAAEQLAKWIADRTDVHIRVFRPPNYSGTIALALLVSLVGGLLYLRRNNLEFIYNKTGWAMVSLCIVFAMTSGQMWNHIRGPPYAHKNPHNGQVSYIHGSSQAQFVAESHIILVLNAAITMGMVLLNEAATSKGDVGKRRIICLVGLGLVVFFFSFLLSIFRSKYHGYPYSFLIK | |
RUO | |
TUSC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7991 | |
TUSC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D8S1992/M33/MGC13453/MRT7/N33/OST3A | |
TUSC3 | |
Recombinant | |
wheat germ expression system |