missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBE2O Partial ORF (NP_071349.2, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | UBE2O (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 37.84kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015615
|
Abnova™
H00063893-Q01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 06-10-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16025615
|
Abnova™
H00063893-Q01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 06-10-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
Sequence: VEPAKIAWECPEKNCAQGEGSMAKKVKRLLKKQVVRIMSCSPDTQCSRDHSMEDPDKKGESKTKSEAESASPEETPDGSASPVEMQDEGAEEPHEAGEQLPPFLLKEGRDSpecifications
UBE2O (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
37.84kDa |