Learn More
Abnova™ Human UGP2 Full-length ORF (NP_006750.3, 1 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007360-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The enzyme encoded by this gene is an important intermediary in mammalian carbohydrate interconversions. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP-glucose and MgPPi. In liver and muscle tissue, UDP-glucose is a direct precursor of glycogen; in lactating mammary gland it is converted to UDP-galactose which is then converted to lactose. The eukaryotic enzyme has no significant sequence similarity to the prokaryotic enzyme. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSRFVQDLSKAMSQDGASQFQEVIRQELELSVKKELEKILTTASSHEFEHTKKDLDGFRKLFHRFLQEKGPSVDWGKIQRPPEDSIQPYEKIKARGLPDNISSVLNKLVVVKLNGGLGTSMGCKGPKSLIGVRNENTFLDLTVQQIEHLNKTYNTDVPLVLMNSFNTDEDTKKILQKYNHCRVKIYTFNQSRYPRINKESLLPVAKDVSYSGENTEAWYPPGHGDIYASFYNSGLLDTFIGEGKEYIFVSNIDNLGATVDLYILNHLMNPPNGKRCEFVMEVTNKTRADVKGGTLTQYEGKLRLVEIAQVPKAHVDEFKSVSKFKIFNTNNLWISLAAVKRLQEQNAIDMEIIVNAKTLDGGLNVIQLETAVGAAIKSFENSLGINVPRSRFLPVKTTSDLLLVMSNLYSLNAGSLTMSEKREFPTVPLVKLGSSFTKVQDYLRRFESIPDMLELDHLTVSGDVTFGKNVSLKGTVIIIANHGDRIDIPPGAVLENKIVSGNLRILDHSpecifications
NP_006750.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UDPG/UDPGP2/UGPP2/pHC379 | |
UGP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7360 | |
UGP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRFVQDLSKAMSQDGASQFQEVIRQELELSVKKELEKILTTASSHEFEHTKKDLDGFRKLFHRFLQEKGPSVDWGKIQRPPEDSIQPYEKIKARGLPDNISSVLNKLVVVKLNGGLGTSMGCKGPKSLIGVRNENTFLDLTVQQIEHLNKTYNTDVPLVLMNSFNTDEDTKKILQKYNHCRVKIYTFNQSRYPRINKESLLPVAKDVSYSGENTEAWYPPGHGDIYASFYNSGLLDTFIGEGKEYIFVSNIDNLGATVDLYILNHLMNPPNGKRCEFVMEVTNKTRADVKGGTLTQYEGKLRLVEIAQVPKAHVDEFKSVSKFKIFNTNNLWISLAAVKRLQEQNAIDMEIIVNAKTLDGGLNVIQLETAVGAAIKSFENSLGINVPRSRFLPVKTTSDLLLVMSNLYSLNAGSLTMSEKREFPTVPLVKLGSSFTKVQDYLRRFESIPDMLELDHLTVSGDVTFGKNVSLKGTVIIIANHGDRIDIPPGAVLENKIVSGNLRILDH | |
RUO | |
UGP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |