Learn More
Abnova™ Human UGT1A10 Partial ORF (NP_061948, 187 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054575-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme encoded by this gene has glucuronidase activity on mycophenolic acid, coumarins, and quinolines. [provided by RefSeq]
Sequence: LSYVPNDLLGFSDAMTFKERVWNHIVHLEDHLFCQYLFRNALEIASEILQTPVTAYDLYSHTSIWLLRTDFVLDYPKPVMPNMIFIGGINCHQGKPLPMEFEASpecifications
NP_061948 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LSYVPNDLLGFSDAMTFKERVWNHIVHLEDHLFCQYLFRNALEIASEILQTPVTAYDLYSHTSIWLLRTDFVLDYPKPVMPNMIFIGGINCHQGKPLPMEFEA | |
RUO | |
UGT1A10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54575 | |
UGT1A10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UDPGT/UGT1J | |
UGT1A10 | |
Recombinant | |
wheat germ expression system |