missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UNC5D Partial ORF (NP_543148, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00137970-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16151887

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: TDNGEALPESIPSAPGTLPHFIEEPDDAYIIKSNPIALRCKARPAMQIFFKCNGEWVHQNEHVSEETLDESSGLKVREVFINVTRQQVEDFHGPEDYWCQ
Specifications

Specifications

NP_543148
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TDNGEALPESIPSAPGTLPHFIEEPDDAYIIKSNPIALRCKARPAMQIFFKCNGEWVHQNEHVSEETLDESSGLKVREVFINVTRQQVEDFHGPEDYWCQ
RUO
UNC5D
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
137970
UNC5D (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16019/KIAA1777/PRO34692/Unc5h4
UNC5D
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers