missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UNC84A Full-length ORF (AAH13613.1, 1 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH13613.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23353 |
Molecular Weight (g/mol) | 54.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177192
|
Abnova™
H00023353-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16167192
|
Abnova™
H00023353-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MDFSRLHMYSPPQCVPENTGYTYALSSSYSSDALDFETEHKLDPVFDSPRMSRRSLRLATTACTLGDGEAVGADSGTSSAVSLKNRAARTTKQRRSTNKSAFSINHVSRQVTSSGVSHGGTVSLQDAVTRRPPVLDESWIREQTTVDHFWGLDDDGDLKGGNKAAIQGNGDVGAAAATAHNGFSCSNCSMLSERKDVLTAHPAPPGPVSRVYSRDRNQKCKSQSFKTQKKVCFPNLIFPFCKSQCLHYLSWRLKIIPSpecifications
AAH13613.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDFSRLHMYSPPQCVPENTGYTYALSSSYSSDALDFETEHKLDPVFDSPRMSRRSLRLATTACTLGDGEAVGADSGTSSAVSLKNRAARTTKQRRSTNKSAFSINHVSRQVTSSGVSHGGTVSLQDAVTRRPPVLDESWIREQTTVDHFWGLDDDGDLKGGNKAAIQGNGDVGAAAATAHNGFSCSNCSMLSERKDVLTAHPAPPGPVSRVYSRDRNQKCKSQSFKTQKKVCFPNLIFPFCKSQCLHYLSWRLKIIP | |
RUO | |
UNC84A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23353 | |
UNC84A (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12407/KIAA0810/MGC176649/SUN1 | |
UNC84A | |
Recombinant | |
wheat germ expression system |