missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UNKL Full-length ORF (AAH00150.1, 1 a.a. - 229 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064718-P02.2ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MTCCSQVPPRRRPSLALSPRLDCNGLNGVPGSIWDFVSGSFSPSPSPILSAGPPSSSSASPNGAELARVRRQLDEAKRKIRQWEESWQQVKQVCDAWQREAQEAKERARVADSDRQLALQKKEEVEAQVKQLQEELEGLGVASTLPGLRGCGDIGTIPLPKLHSLQSQLRLDLEAVDGVIFQLRAKQCVACRERAHGAVLRPCQHHILCEPCAATAPECPYCKGQPLQWSpecifications
AAH00150.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C16orf28/FLJ12623/FLJ23360/KIAA0734/MGC5179/ZC3H5L/ZC3HDC5L | |
UNKL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64718 | |
UNKL (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTCCSQVPPRRRPSLALSPRLDCNGLNGVPGSIWDFVSGSFSPSPSPILSAGPPSSSSASPNGAELARVRRQLDEAKRKIRQWEESWQQVKQVCDAWQREAQEAKERARVADSDRQLALQKKEEVEAQVKQLQEELEGLGVASTLPGLRGCGDIGTIPLPKLHSLQSQLRLDLEAVDGVIFQLRAKQCVACRERAHGAVLRPCQHHILCEPCAATAPECPYCKGQPLQW | |
RUO | |
UNKL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |