Learn More
Abnova™ Human USF2 Partial ORF (NP_003358, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007392-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MDMLDPGLDPAASATAAAAASHDKGPEAEEGVELQEGGDGPGAEEQTAVAITSVQQAAFGDHNIQYQFRTETNGGQVTYRVVQVTDGQLDGQGDTAGAVSSpecifications
NP_003358 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDMLDPGLDPAASATAAAAASHDKGPEAEEGVELQEGGDGPGAEEQTAVAITSVQQAAFGDHNIQYQFRTETNGGQVTYRVVQVTDGQLDGQGDTAGAVS | |
RUO | |
USF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7392 | |
USF2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FIP/bHLHb12 | |
USF2 | |
Recombinant | |
wheat germ expression system |